Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Manometers
Pellicon® Gauges
Pellicon® Gauges
Pellicon® Gauges
Catalog # MILLXX4200075
Supplier:
Merck
Expand image
Pellicon® Gauges
Catalog # MILLXX4200075
Supplier:
Merck
Specifications
Range:
0 - 7 bar
Description:
1/4" NPTM gauge, made of stainless steel
Pk:
1
Specifications
About this item
Product Details
Product Details