Order Entry
France
ContactUsLinkComponent
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% (by titrimetric analysis), cGMP-manufactured
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% (by titrimetric analysis), cGMP-manufactured
Supplier:  Spectrum Chemical
Certificates
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% (by titrimetric analysis), cGMP-manufactured
Supplier:  Spectrum Chemical

Specification Test Results

  • APPEARANCE:
    WHITE FINE POWDER
  • ASSAY (TITRATION):
    min. 99.5 %
  • pH OF A 5% AQUEOUS SOLUTION:
    4.5 - 6.5
  • pKa @ 25°C:
    7.35- 7.75
  • Absorbance @ 260 nm (0.05 M in water):
    max. 0.05 AU
  • Absorbance @ 280 nm (0.05 M in water):
    max. 0.03 AU
  • WATER (KF):
    max. 0.5 %
  • HEAVY METALS (as Pb) :
    max. 5 ppm
  • INSOLUBLE MATTER :
    max. 0.1 %
  • TOTAL AEROBIC MICROBIAL COUNT :
    max. 100 cfu/g
  • TOTAL YEAST AND MOLD COUNT :
    max. 100 cfu/g
  • BACTERIAL ENDOTOXIN:
    As reported

Specification Test Results

Frequently Bought Together