Order Entry
France
ContactUsLinkComponent
VWR® 32-Place PCR Racks
VWR® 32-Place PCR Racks
Catalog # 216-2988
Supplier:  VWR Collection
undefined
VWR® 32-Place PCR Racks
Catalog # 216-2988
Supplier:  VWR Collection

Specifications

  • Colour:
    Blue
  • Description:
    VWR® collection 32-place PCR rack, hold 2 ml tubes
  • Material:
    PP
  • W×D×H:
    175×98×31 mm
  • Pk:
    10

Specifications

About this item

Designed for pre- and post-PCR sampling, streamline your PCR workflow with the 32-Place PCR rack. This versatile rack accommodates 0,2 ml tubes both individually and in strips of 8, as well as 32-place assay plates (8×4).

  • Small and compact design is stackable, saving valuable space in the freezer (Low temperature range: −80 °C)
  • Right sized when processing smaller batches
  • Lid hinge design allows lid to sit at 180 degrees, clear of sample processing, or be removed entirely
  • Transparent lid protects samples and allows quick assessment of contents
  • Embossed numbers and letters on the wells provide easy identification and tracking of samples
  • Light-weight construction made durable for reusability
  • Autoclavable

Capacity: 32-Place
Material: 100% Recyclable polypropylene (PP-05)