Order Entry
France
ContactUsLinkComponent
Depyrogenated Dilution Tubes
Depyrogenated Dilution Tubes
Catalog # WAKFDL-13100
CAS Number:  
undefined
Depyrogenated Dilution Tubes
Catalog # WAKFDL-13100
CAS Number:  

Specifications

  • Pk:
    50
  • Description:
    Depyrogenated dilution tubes, 13×100 mm, <0,001 EU/tube, 50 tubes, borosilicate glass

Specifications

About this item

These depyrogenated dilution tubes are made of high-quality borosilicate glass depyrogenated at 250 °C.

  • Endotoxin-free test tubes (<0,001 EU/tube)
  • Designed for all testing methods
  • Made with borosilicate glass
  • Can be used for general lab testing

This results in endotoxin-free tubes that can perform successful LAL dilution series, from creating the standard curve to sample dilutions, with minimal chance of potential contamination.