ISAspher HPLC Columns, Amino
: ISERA
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
- The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
- Reliable and proven
- Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µm particle size) can be easily implemented. Of course, the same applies for a scale-down, e.g. to a 3 µm particle size, if a speed-optimised analytical method has to be established.
: Individual test certificate is enclosed with each column.
: Columns are delivered in solvent which is suitable for normal phase chromatography.
- Catalog No:
- ISERA111-325U00
- ISERA111-325W00
- ISERA111-325V00
- ISERA111-323B00
- ISERA111-323A00
- ISERA111-325R00
- ISERA111-325Q00
- ISERA111-325T00
- ISERA111-325S00
- ISERA111-325M00
- ISERA111-325L00
- ISERA111-325P00
- ISERA111-325N00
- ISERA111-325K00
- ISERA111-325J00
- ISERA111-325H00
- ISERA111-325E00
- ISERA111-325D00
- ISERA111-329Z00
- ISERA111-325G00
- ISERA111-325F00
- ISERA111-325A00
- ISERA111-329W00
- ISERA111-323Z00
- ISERA111-329V00
- ISERA111-325C00
- ISERA111-329Y00
- ISERA111-325B00
- ISERA111-329X00
- ISERA111-329S00
- ISERA111-329R00
- ISERA111-323Y00
- ISERA111-329U00
- ISERA111-329T00
- ISERA111-329Q00
- ISERA111-323X00
- ISERA111-323W00
- ISERA111-323T00
- ISERA111-329P00
- ISERA111-323S00
- ISERA111-329N00
- ISERA111-323V00
- ISERA111-323U00
- ISERA111-323P00
- ISERA111-329K00
- ISERA111-323N00
- ISERA111-329J00
- ISERA111-323R00
- ISERA111-329M00
- ISERA111-323Q00
- ISERA111-329L00
- ISERA111-329F00
- ISERA111-329E00
- ISERA111-329H00
- ISERA111-329G00
- ISERA111-323L00
- ISERA111-323K00
- ISERA111-323M00
- ISERA111-329C00
- ISERA111-323G00
- ISERA111-329B00
- ISERA111-323F00
- ISERA111-323J00
- ISERA111-323H00
- ISERA111-329D00
- ISERA111-323C00
- ISERA111-325Y00
- ISERA111-325X00
- ISERA111-329A00
- ISERA111-323E00
- ISERA111-325Z00
- ISERA111-323D00
- Phase name:
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- Carbon load:
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- pH range:
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- Surface area:
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- USP code:
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- Storage:
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- Shipping conditions:
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
Frequently Bought Together