Order Entry
France
ContactUsLinkComponent
Calcium-D-saccharate tetrahydrate ≥98.5% USP
Irritant
Calcium-D-saccharate tetrahydrate ≥98.5% USP
Catalog # SPCMC1830-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Calcium-D-saccharate tetrahydrate ≥98.5% USP
Catalog # SPCMC1830-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    98.5 - 102.0%
  • Specific Rotation [A]D:
    +18.5 to +22.5°
  • Chloride (Cl):
    max. 0.07%
  • Sulfate:
    max. 0.12%
  • Heavy Metals:
    max. 0.002%

Specification Test Results

About this item

 :
 :320.26 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:5793-89-5
Formula:C₆H₈CaO₈.4H₂O
MDL Number:MFCD00150704