Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 FCC
Catalog # SPCMP1216-500GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly pail
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:9003-39-8
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626