Order Entry
France
ContactUsLinkComponent
Aluminium potassium sulphate dodecahydrate 99.5-100.5%, crystals FCC
Aluminium potassium sulphate dodecahydrate 99.5-100.5%, crystals FCC
Catalog # SPCMA1107-12KG
Supplier:  Spectrum Chemical
CAS Number:  
Aluminium potassium sulphate dodecahydrate 99.5-100.5%, crystals FCC
Catalog # SPCMA1107-12KG
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Assay [Alk(SO₄)₂[-12H₂O]:
    99.5-100.5%
  • Ammonium Salts:
    To Pass Test
  • Fluoride:
    max. 0.003%
  • Lead (Pb):
    max. 3 mg/kg
  • Selenium (Se):
    max. 0.003%

Specification Test Results

About this item

 :
 :474.39 g/mol
Melting Pt:92,5 °C/198,5 °F
Density:1,757
Storage Temperature:Ambient
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143