Order Entry
France
ContactUsLinkComponent
AMP (2-Amino-2-methylpropanol)
Irritant
AMP (2-Amino-2-methylpropanol)
Catalog # SPCMAM156-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
AMP (2-Amino-2-methylpropanol)
Catalog # SPCMAM156-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    200 L
  • Seq:
    5549

Specifications

About this item

 :
Merck Index:13,00448
Boiling Pt:165 °C (1013 hPa)
Melting Pt:18…26 °C
Density:0,934
Flash Pt:67 °C
 :89.14 g/mol
Storage Temperature:Ambient
EINECS:204-709-8
 :GHS07
CAS Number:124-68-5
Formula:(CH₃)₂C(NH₂)CH₂OH
MDL Number:MFCD00008051