Anti-Claudin 16 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BOSSBS-13752R-HRP
: Bioss
- Antibody type:Primary
- Antigen name:Claudin 16
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Epitope:95-150/305
- Form:Liquid
- Gene ID:10686
- Antigen synonyms:HOMG3|PCLN-1|Paracellin-1|Claudin-16|CLDN16|PCLN1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:34 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Claudin 16
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity. Involved in paracellular magnesium reabsorption. Required for a selective paracellular conductance. May form, alone or in partnership with other constituents, an intercellular pore permitting paracellular passage of magnesium and calcium ions down their electrochemical gradients. Alternatively, it could be a sensor of magnesium concentration that could alter paracellular permeability mediated by other factors.
Type: Primary
Antigen: Claudin 16
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 95-150/305
Host: Rabbit
Isotype: IgG
Reactivity: