Anti-WDR61 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-6886R-A680
: Bioss
- Antibody type:Primary
- Antigen name:WDR61
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Epitope:121-220/305
- Form:Liquid
- Gene ID:80349
- Antigen synonyms:WD repeat containing protein 61|WD repeat domain 61|WDR61_HUMAN|Meiotic recombination REC14 protein homolog|SKI8|WDR 61|Recombination protein REC14|REC14
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human WDR61
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
WDR61 is a subunit of the human PAF and SKI complexes, which function in transcriptional regulation and are involved in events downstream of RNA synthesis, such as RNA surveillance (Zhu et al., 2005 [PubMed 16024656]).[supplied by OMIM, Mar 2008].
Type: Primary
Antigen: WDR61
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 121-220/305
Host: Rabbit
Isotype: IgG
Reactivity: Rat