Anti-TCF3 Thr355 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-6066R-A750
: Bioss
- Antibody type:Primary
- Antigen name:TCF3 Thr355
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:301-400/651
- Form:Liquid
- Gene ID:6929
- Antigen synonyms:VDIR|E12|Immunoglobulin enhancer-binding factor E12/E47|MGC129647|TCF3/E2Aphospho T355|TFE2_MOUSE|Transcription factor ITF-1|TCF-3|TFE2|MGC129648|p-TCF3phospho T355|Helix loop helix protein HE47|bHLHb21|VDR interacting repressor|TCF3|Kappa-E2-binding factor|Class B basic helix-loop-helix protein 21|ITF1|Negative vitamin D response element binding protein|transcription factor 3|E47|E2A|immunoglobulin transcription factor 1|Kappa E2 binding factor|E2A immunoglobulin enhancer binding factors E12/E47|Transcription factor E2-alpha|p-E47phospho T355
- Modification:Phosphorylation (Thr355)
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:67 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human TCF3 around the phosphorylation site of Thr355
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Heterodimers between TCF3 and tissue-specific basic helix-loop-helix (bHLH) proteins play major roles in determining tissue-specific cell fate during embryogenesis, like muscle or early B-cell differentiation. Dimers bind DNA on E-box motifs: 5'-CANNTG-3'. Binds to the kappa-E2 site in the kappa immunoglobulin gene enhancer.
Type: Primary
Antigen: TCF3 Thr355
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-400/651
Host: Rabbit
Isotype: IgG
Reactivity: