- Antibody type:Primary
- Antigen name:BLVRA/Biliverdin Reductase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Epitope:161-260/296
- Form:Liquid
- Gene ID:644
- Antigen synonyms:BVR|BLVRA|BVR A|BLVR|BVRA|Biliverdin reductase A|BIEA_HUMAN|zinc-metalloprotein|BLVR A|Biliverdin IX alpha reductase
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BLVRA/Biliverdin Reductase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
In human liver cytosolic fractions, four forms of biliverdin reductase have been identified, including two biliverdin-IX Beta reductases and two biliverdin-IX Alpha reductases, designated isozymes I and II and isozymes III and IV, respectively. Biliverdin reductase A (BLVRA), also designated biliverdin-IX Alpha-reductase, belongs to the GFO/iIDH/MocA family and the biliverdin reductase subfamily. The gene that encodes this cytoplasmic protein maps to chromosome 7p14-cen. BLVRA reduces biliverdin IX (the methene bridge of the open tetrapyrrole) to bilirubin with the concomitant oxidation of an NADH or NADPH cofactor (bilirubin + NADP+ = biliverdin + NADPH). BLVRA is expressed primarily in liver.
Type: Primary
Antigen: BLVRA/Biliverdin Reductase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 161-260/296
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat