Order Entry
France
ContactUsLinkComponent
Anti-APPBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-APPBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-12503R-A680
Supplier:  Bioss
Anti-APPBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-12503R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    APPBP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    401-500/534
  • Form:
    Liquid
  • Gene ID:
    8883
  • Antigen synonyms:
    NAE1|Amyloid beta precursor protein-binding protein 1, 59 kDa|APPBP1|Proto-oncogene protein 1|A-116A10.1|NEDD8-activating enzyme E1 regulatory subunit|ula-1|APP-BP1|HPP1|Amyloid protein-binding protein 1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    60 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human APPBP1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Regulatory subunit of the dimeric UBA3-NAE1 E1 enzyme. E1 activates NEDD8 by first adenylating its C-terminal glycine residue with ATP, thereafter linking this residue to the side chain of the catalytic cysteine, yielding a NEDD8-UBA3 thioester and free AMP. E1 finally transfers NEDD8 to the catalytic cysteine of UBE2M. Necessary for cell cycle progression through the S-M checkpoint. Overexpression of NAE1 causes apoptosis through deregulation of NEDD8 conjugation.

Type: Primary
Antigen: APPBP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-500/534
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat