Order Entry
France
ContactUsLinkComponent
Anti-Doppel/DPL Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Doppel/DPL Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11732R-A750
Supplier:  Bioss
Anti-Doppel/DPL Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11732R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Doppel/DPL
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    51-120/176
  • Form:
    Liquid
  • Antigen synonyms:
    PRND|Prion protein 2|PRND_HUMAN|Prion protein 2 dublet|PrPLP|MGC41841|Prion gene complex downstream|Dublet|Prion-like protein doppel|Prion like protein doppel|DPL
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    14 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Doppel
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Prion diseases or transmissible spongiform encephalopathies (TSEs) are manifested as genetic, infectious or sporadic, lethal neurodegenerative disorders involving alterations of the prion protein (PrP). Infectious PrPSc is highly expressed in the brain of animals affected by TSEs, including scrapie in sheep, BSE in cattle, and Cruetzfeldt-Jacob disease in humans. The PRND gene locus, located on human chromosome 20p, encodes for the doppel protein (Dpl), which exhibits approximately 25% sequence homology with PrP. Dpl is characterised by an alpha-helical conformation, intramolecular disulfide bonds, and two N-linked oligosaccharides, and it is presented on the cell surface by a glycosylphosphatidylinositol anchor. Dpl is highly expressed in adult testis and heart and is detectable in the brain of neonatal mice. Dpl does not appear to contribute to prion disease progression, but ectopic expression of Dpl is implicated in neuronal degeneration of ataxic PRP-deficient mice. Dpl is also thought to play a role in angiogenesis, specifically maturation of the blood-brain barrier.

Type: Primary
Antigen: Doppel/DPL
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-120/176
Host: Rabbit
Isotype: IgG
Reactivity: