Order Entry
France
ContactUsLinkComponent
Anti-PI3 Kinase p110 delta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-PI3 Kinase p110 delta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-10656R-A750
 :  Bioss
Anti-PI3 Kinase p110 delta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-10656R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    PI3 Kinase p110 delta
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    51-150/1044
  • Form:
    Liquid
  • Gene ID:
    5293
  • Antigen synonyms:
    Phosphatidylinositol 3 kinase catalytic delta polypeptide|Phosphatidylinositol 45 bisphosphate 3 kinase catalytic subunit delta isoform|p110delta|p110D|Phosphatidylinositol 4 5 bisphosphate 3 kinase catalytic subunit delta isoform
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    115 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human PI3 Kinase p110 delta
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Phosphoinositide-3-kinase (PI3K) that phosphorylates PftdIns (4,5)P2 (Phosphatidylinositol 4,5-bisphosphate) to generate phosphatidylinositol 3,4,5-trisphosphate (PIP3). PIP3 plays a key role by recruiting PH domain-containing proteins to the membrane, including AKT1 and PDPK1, activating Signalling cascades involved in cell growth, survival, proliferation, motility and morphology. Mediates immune responses. Plays a role in B-cell development, proliferation, migration, and function. Required for B-cell receptor (BCR) Signalling. Mediates B-cell proliferation response to anti-IgM, anti-CD40 and IL4 stimulation. Promotes cytokine production in response to TLR4 and TLR9. Required for antibody class switch mediated by TLR9. Involved in the antigen presentation function of B-cells. Involved in B-cell chemotaxis in response to CXCL13 and sphingosine 1-phosphate (S1P). Required for proliferation, Signalling and cytokine production of naive, effector and memory T-cells. Required for T-cell receptor (TCR) Signalling. Mediates TCR Signalling events at the immune synapse. Activation by TCR leads to antigen-dependent memory T-cell migration and retention to antigenic tissues. Together with PIK3CG participates in T-cell development. Contributes to T-helper cell expansion and differentiation. Required for T-cell migration mediated by homing receptors SELL/CD62L, CCR7 and S1PR1 and antigen dependent recruitment of T-cells. Together with PIK3CG is involved in natural killer (NK) cell development and migration towards the sites of inflammation. Participates in NK cell receptor activation. Have a role in NK cell maturation and cytokine production. Together with PIK3CG is involved in neutrophil chemotaxis and extravasation. Together with PIK3CG participates in neutrophil respiratory burst. Have important roles in mast-cell development and mast cell mediated allergic response. Involved in stem cell factor (SCF)-mediated proliferation, adhesion and migration.

Type: Primary
Antigen: PI3 Kinase p110 delta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/1044
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse