- Antibody type:Primary
- Antigen name:beta Adaptin/AP2 beta
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:351-450/937
- Form:Liquid
- Gene ID:2971
- Antigen synonyms:Plasma membrane adaptor HA2/AP2 adaptin beta subunit|AP 105A|AP2B1|_-Adaptin|AP1 complex subunit beta 1|ADTB 2|Clathrin assembly protein complex 1|Plasma membrane adaptor HA2/AP2 adaptor beta subunit|Adaptin beta 2|CLAPB 2|Adaptor related protein complex 2, beta 1 subunit|AP2beta|Clathrin assembly protein complex 1 beta large chain|Adaptin beta 2 beta|CLAPB 1|AP 105B|DKFZp781K0743|AP1B1|Adaptor related protein complex 1 beta 1 subunit|Golgi adaptor HA1/AP1 adaptin beta subunit|AP2 beta|AP105B|ADTB 1|CLAPB2|BAM22|Adaptin, beta 2 beta|Adaptor related protein complex 1, beta 1 subunit|AP 1 complex subunit beta 1|Clathrin assembly protein complex 2 beta large chain|AP105A|BAM 22|ADTB1|AP 2 complex subunit beta 1|Adaptin beta 1 beta prime|Clathrin associated/assembly/adaptor protein large beta 1|ADTB2|Adaptor related protein complex 2 beta 1 subunit|Adaptin-_1|AP1 Beta|Beta adaptin|Beta1 adaptin|CLAPB1|Beta prime adaptin|Beta2 adaptin|Plasma membrane adaptor HA2/AP2.|Adaptor protein complex AP 1 beta 1 subunit|Adaptin _1|AP2 complex subunit beta 1|Clathrin associated/assembly/adaptor protein large beta 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:103 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human beta Adaptin
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limbs and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. AP2 beta appears to be required for normal face and limb development and for proper terminal differentiation and function of renal tubular epithelia.
Type: Primary
Antigen: beta Adaptin/AP2 beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 351-450/937
Host: Rabbit
Isotype: IgG
Reactivity: