Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2653R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SPHK2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:101-200/654
- Form:Liquid
- Gene ID:56848
- Antigen synonyms:SK-2|SK 2|SPHK2|Sphingosine kinase 2|SPK-2|SPK 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:72 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SPHK2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Catalyses the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-dihydrosphingosine, D-erythro-sphingosine and L-threo-dihydrosphingosine. Binds phosphoinositides.
Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/654
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat