Anti-Aldolase A, Aldolase B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-2406R-A750
: Bioss
- Antibody type:Primary
- Antigen name:Aldolase A, Aldolase B
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:261-364/364
- Form:Liquid
- Gene ID:226
- Antigen synonyms:ALDA|Muscle-type aldolase|Fructose-bisphosphate aldolase A|Lung cancer antigen NY-LU-1|GSD12|HEL-S-87p|ALDOA
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Aldolase A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Plays a key role in glycolysis and gluconeogenesis. In addition, may also function as scaffolding protein (By similarity).
Type: Primary
Antigen: Aldolase A, Aldolase B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 261-364/364
Host: Rabbit
Isotype: IgG
Reactivity: Mouse