- Antibody type:Primary
- Antigen name:TCAB1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:285-390/548
- Form:Liquid
- Gene ID:3135
- Antigen synonyms:FLJ10385|TCAB1|WAP53_HUMAN|WDR79/TCAB1|telomerase Cajal Body protein 1|WD repeat containing protein 79|WRAP53
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:60 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human WDR79/TCAB1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
WDR79 contains six WD (tryptophan-aspartate) repeat domains found in a number of proteins that function as adaptor molecules in signal transduction and cytoskeletal organization. The WD repeat is defined by four or more repeating units of a conserved core of approximately 40 amino acids ending with tryptophan-aspartic acid (WD). WD repeats may serve as sites of protein-protein interaction for adaptor proteins and facilitate multiprotein complex formation. The function of the WDR79 protein has not been characterized, however significant and consistent single nucleotide polymorphisms in the WDR79 gene have been found to be associated with ER negative breast cancer.
Type: Primary
Antigen: TCAB1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 285-390/548
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat