- Pk:96 Tests
- Assay Type:Competitive
- Host:
- Primary antibody reactivity:
- Description:QuickDetect™ 8-OHdG (Rat) ELISA Kit, 100 assays
- Environmentally Preferable:
A rapid sandwich ELISA kit for quantify 8-OHdG in rat serum, plasma, urine and other biological fluid.
- Easy, convenient and time-saving method to measure the level of 8-OHdG in rat serum, plasma, urine, cell culture samples, tissue samples
- Detection Range: 22 to 1800 pg/ml
- Sensitivity: <5 pg/ml
8-hydroxy-2'-deoxyguanosine (8-OHdG) is an oxidised derivative of deoxyguanosine. Concentrations of 8-oxo-dG within a cell are a measurement of oxidative stress. It is considered as a critical biomarker of oxidative stress and carcinogenesis. This ELISA kit uses Competitive-ELISA as the method. The microtiter plate provided in this kit has been pre-coated with 8-OHdG. During the reaction,8-OHdG in the sample or standard competes with a fixed amount of 8-OHdG on the solid phase supporter for sites on the Biotinylated Detection Antibody specific to 8-OHdG. BioVision’s 8-OHdG ELISA kit is a sandwich ELISA assay for the quantitative measurement of Rat 8-OHdG in serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of Rat 8-OHdG captured from the samples.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.