Order Entry
France
ContactUsLinkComponent
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Flammable
Health
Irritant
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Catalog # 1.01774.6025
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Catalog # 1.01774.6025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Stainless steel drum
  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Solubility:
    (25 °C) practically insoluble
  • Boiling Point:
    60 - 80 °C
  • Vapor Pressure:
    200 hPa (20 °C)
  • Flash Point:
    -26 °C
  • Explosion Limit:
    1.0 - 7.4 %(V)
  • Density:
    0.68 g/cm3 (20 °C)
  • Ignition Point:
    240 °C

Specification Test Results

About this item

 :
Boiling Pt:60…80 °C (1013 hPa)
Density:0.67 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849