Magnesium aluminum silicate NF Type IA
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:1327-43-1
:
Storage Temperature:Ambient
Specification Test Results
- Viscosity @ 33 +/- 3 °C:225 - 600 cP
- pH of a 5 in 100 Suspension:9.0 - 10.0
- Ratio of Content Aluminum / Magnesium:0.5 - 1.2
- Acid Demand:max. 4.0
- Loss On Drying:max. 8.0%
Specification Test Results
Frequently Bought Together