Pyridoxine hydrochloride ≥98.0% USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:58-56-0
:
Formula:C₈H₁₂ClNO₃
MDL Number:MFCD00012807
:205.64 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (C₈H₁₁NO₃[-HCl):98.0 - 102.0 %
- Chloride Content:16.9 - 17.6 %
- Loss On Drying:max. 0.5 %
- Residue After Ignition:max. 0.1 %
- Heavy Metals:max. 0.003 %
Specification Test Results
Frequently Bought Together