Order Entry
France
Orders LinkContactUsLinkComponent
Elbows
Elbows
Supplier:   LABCONCO
Elbows
Supplier:  LABCONCO
Certificates
3 Options Available Below
Product Details & Documents
PVC elbows 90° are compatible with thermoplastic duct.
Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.
Product Family Options
Product Information
  • Availability
    Price
  • LBCP4708700
    90° elbows, Ø 152 mm, Height - 343 mm
    90° elbows, Ø 152 mm, Height - 343 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce
  • LBCP7027300
    90° elbows, Ø 254 mm, Height - 508 mm
    90° elbows, Ø 254 mm, Height - 508 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce
  • LBCP5602100
    90° elbows, Ø 305 mm, Height - 610 mm
    90° elbows, Ø 305 mm, Height - 610 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce