Toluene, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. AH
Catalog # HONC65752-195L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Stainless steel drum, returnable
- Pk:195 L
- Seq:5531
Specifications
About this item
:
:92.14 g/mol
Storage Temperature:Ambient
UN:1294
ADR:3,II
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512