Order Entry
France
ContactUsLinkComponent
Accessories for boekel tube rotator
Accessories for boekel tube rotator
Supplier:  BOEKEL SCIENTIFIC
Certificates
Accessories for boekel tube rotator
Supplier:  BOEKEL SCIENTIFIC
Specifications
  • Catalog No:
  • BOEI260651
  • BOEI260653
  • BOEI260652
  • Capacity:
  • 27-34 mm tubes
  • Speed:
  • 4-80 min⁻¹
  • Angle:
  • 90 - 180°
  • W×D×H:
  • 304×431×381 mm
  • Electrical:
  • 19,4 vdc, 4,74 A via 100 - 240 AC
  • Timer:
  • Up to 99 hrs. and 59 min.