Prothioconazole Analytical standard, Fluka™
Catalog # HONCR1935-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:178928-70-6
- Description:Prothioconazole Analytical standard
- Storage conditions:RT
- Synonym(s):2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione
- Melting point:140.3 °C
- Formula:C₁₄H₁₅Cl₂N₃OS
- Molecular weight:344.26 g/mol
- Description:Prothioconazole
- Physical form:Crystalline
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery