Repaglinide Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- 1600824.
- 1600835.
- 1600846.
- 1600868.
- Description:
- Repaglinide Related Compound A
- Repaglinide Related Compound B
- Repaglinide Related Compound C
- Repaglinide Related Compound E
- Synonym(s):
- (S)-3-Methyl-1-[2-(1-piperidinyl)phenyl]butylamine, N-acetyl-L-glutamate salt
- 3-Ethoxy-4-ethoxycarbonylphenylacetic acid
- (S)-2-Ethoxy-4-[2-[[2-phenyl-1-[2-(1-piperidinyl)phenyl]ethyl]amino]-2-oxoethyl] benzoic acid
- (R)-2-Ethoxy-4-[2-({3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl}amino)-2-oxoethyl]benzoic acid
- CAS No.:
- 219921-94-5
- 99469-99-5
- 1803084-79-8
- 147852-26-4
- Formula:
- C₂₃H₃₇N₃O₅
- C₁₃H₁₆O₅
- C₂₇H₃₆N₂O₄
- UN No.:
- 2811
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1600824.Repaglinide Related Compound ARepaglinide Related Compound AUSP reference standardVial10 mgLog in for availability1 * 10 mg1 170,00 €Specifications:
More
Specifications
Cat. No.1600824.DescriptionRepaglinide Related Compound ASynonym(s)(S)-3-Methyl-1-[2-(1-piperidinyl)phenyl]butylamine, N-acetyl-L-glutamate saltCAS No.219921-94-5FormulaC₂₃H₃₇N₃O₅UN No. - 1600835.Repaglinide Related Compound BRepaglinide Related Compound BUSP reference standardVial50 mgLog in for availability1 * 50 mg1 100,00 €Specifications:
More
Specifications
Cat. No.1600835.DescriptionRepaglinide Related Compound BSynonym(s)3-Ethoxy-4-ethoxycarbonylphenylacetic acidCAS No.99469-99-5FormulaC₁₃H₁₆O₅UN No. - 1600846.Repaglinide Related Compound CRepaglinide Related Compound CUSP reference standardVial25 mgLog in for availability1 * 25 mg1 230,00 €Specifications:
More
Specifications
Cat. No.1600846.DescriptionRepaglinide Related Compound CSynonym(s)(S)-2-Ethoxy-4-[2-[[2-phenyl-1-[2-(1-piperidinyl)phenyl]ethyl]amino]-2-oxoethyl] benzoic acidCAS No.1803084-79-8FormulaUN No.2811 - 1600868.Repaglinide Related Compound ERepaglinide Related Compound EUSP reference standardVial20 mgLog in for availability1 * 20 mg2 240,00 €Specifications:
More
Specifications
Cat. No.1600868.DescriptionRepaglinide Related Compound ESynonym(s)(R)-2-Ethoxy-4-[2-({3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl}amino)-2-oxoethyl]benzoic acidCAS No.147852-26-4FormulaC₂₇H₃₆N₂O₄UN No.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



