Order Entry
France
ContactUsLinkComponent
Metoprolol Fumarate, Reference Standard
Metoprolol Fumarate, Reference Standard
Catalog # 1441287.
Supplier:  USP
CAS Number:  
Metoprolol Fumarate, Reference Standard
Catalog # 1441287.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • CAS No.:
    119637-66-0
  • Description:
    Metoprolol Fumarate
  • Seq:
    05
  • Formula:
    C₁₉H₂₉NO₇
  • Description:
    Metoprolol Fumarate

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard