Order Entry
France
Orders LinkContactUsLinkComponent
 

Decitabine Related Compounds, Reference Standard

 :   USP
Decitabine Related Compounds, Reference Standard
 :  USP
 
Decitabine Related Compounds, Reference Standard

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1165215.
  • 1165226.
  • 1165248.
  • 1165259.
  • Description:
  • Decitabine Related Compound A
  • Decitabine Related Compounds B and C Mixture
  • Decitabine Related Compound D
  • Decitabine Related Compound E
  • Synonym(s):
  • 1-O-Acetyl-3,5-di-O-(4-chlorobenzoyl)-2-deoxy-D-ribofuranose
  • 4-Amino-1-(3,5-di-O-(4-chlorobenzoyl)-2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one and 4-Amino-1-(3,5-di-O-(4-chlorobenzoyl)-2-deoxy-a-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one
  • Methyl 4-chlorobenzoate
  • 4-Amino-1-(2-deoxy-α-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one
  • CAS No.:
  • 1207459-15-1
  • 1126-46-1
  • 22432-95-7

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Decitabine Related Compound A
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 1 260,00 €
    1 260,00 €
    1 * 25 mg
       1 260,00 €

     
     
    Specifications
    Cat. No.
    1165215.
    Description
    Decitabine Related Compound A
    Synonym(s)
    1-O-Acetyl-3,5-di-O-(4-chlorobenzoyl)-2-deoxy-D-ribofuranose
    CAS No.
    1207459-15-1
  • Decitabine Related Compounds B and C Mixture
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 1 260,00 €
    1 260,00 €
    1 * 25 mg
       1 260,00 €

     
     
    Specifications
    Cat. No.
    1165226.
    Description
    Decitabine Related Compounds B and C Mixture
    Synonym(s)
    4-Amino-1-(3,5-di-O-(4-chlorobenzoyl)-2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one and 4-Amino-1-(3,5-di-O-(4-chlorobenzoyl)-2-deoxy-a-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one
    CAS No.
  • Decitabine Related Compound D
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 1 100,00 €
    1 100,00 €
    1 * 25 mg
       1 100,00 €

     
     
    Specifications
    Cat. No.
    1165248.
    Description
    Decitabine Related Compound D
    Synonym(s)
    Methyl 4-chlorobenzoate
    CAS No.
    1126-46-1
  • Decitabine Related Compound E
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 1 240,00 €
    1 240,00 €
    1 * 25 mg
       1 240,00 €

     
     
    Specifications
    Cat. No.
    1165259.
    Description
    Decitabine Related Compound E
    Synonym(s)
    4-Amino-1-(2-deoxy-α-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one
    CAS No.
    22432-95-7
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.