Order Entry
France
Orders LinkContactUsLinkComponent
Bendamustine Related Compound, Reference Standard

Bendamustine Related Compound, Reference Standard

Supplier:   USP
Bendamustine Related Compound, Reference Standard
Supplier:  USP
Certificates
Bendamustine Related Compound, Reference Standard

8 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1065232.
  • 1065301.
  • 1065243.
  • 1065254.
  • 1065265.
  • 1065276.
  • 1065298.
  • 1065312.
  • Description:
  • Bendamustine Related Compound A
  • Bendamustine Related Compound H
  • Bendamustine Related Compound B
  • Bendamustine Related Compound C
  • Bendamustine Related Compound D
  • Bendamustine Related Compound E
  • Bendamustine Related Compound G
  • Bendamustine Related Compound I
  • Synonym(s):
  • 4-{5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoic acid hydrochloride
  • 4-[5-({2-[(4-{5-[Bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoyl)oxy]ethyl}(2-chloroethyl)amino)-1-methyl-1H-benzimidazol-2-yl]butanoic acid
  • 4-(1-Methyl-5-morpholino-1H-benzimidazol-2-yl)butanoic acid hydrochloride
  • Ethyl 4-{5-[bis(2-hydroxyethyl)amino]-1-methyl-1H -benzimidazol-2-yl}butanoate
  • 4-{5-[(2-Chloroethyl)amino]-1-methyl-1H -benzimidazol-2-yl}butanoic acid hydrochloride
  • 4-{5-[(2-Chloroethyl)(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoic acid
  • 4-[6-(2-Chloroethyl)-3,6,7,8-tetrahydro-3-methylimidazo[4,5-h][1,4]benzothiazin-2-yl]butanoic acid hydrochloride
  • Ethyl 4-{5-[bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoate
  • CAS No.:
  • 109882-29-3
  • 1228551-91-4
  • 3543-74-6
  • 1797881-48-1
  • 109882-27-1
  • 87475-54-5
  • Formula:
  • C₃₂H₄₁Cl₃N₆O₄
  • C₁₆H₂₁N₃O₃,xHCl
  • C₁₄H₁₉Cl₂N₃O₂
  • C₁₆H₂₂ClN₃O₃
  • C₁₈H₂₅Cl₂N₃O₂

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1065232.
    Bendamustine Related Compound A
    Bendamustine Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 290,00 €
    1 290,00 €
    1 * 25 mg
       1 290,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1065232.
    Description
    Bendamustine Related Compound A
    Synonym(s)
    4-{5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoic acid hydrochloride
    CAS No.
    109882-29-3
    Formula
  • 1065301.
    Bendamustine Related Compound H
    Bendamustine Related Compound H
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 140,00 €
    1 140,00 €
    1 * 25 mg
       1 140,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1065301.
    Description
    Bendamustine Related Compound H
    Synonym(s)
    4-[5-({2-[(4-{5-[Bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoyl)oxy]ethyl}(2-chloroethyl)amino)-1-methyl-1H-benzimidazol-2-yl]butanoic acid
    CAS No.
    1228551-91-4
    Formula
    C₃₂H₄₁Cl₃N₆O₄
  • 1065243.
    Bendamustine Related Compound B
    Bendamustine Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 390,00 €
    1 390,00 €
    1 * 20 mg
       1 390,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1065243.
    Description
    Bendamustine Related Compound B
    Synonym(s)
    4-(1-Methyl-5-morpholino-1H-benzimidazol-2-yl)butanoic acid hydrochloride
    CAS No.
    Formula
    C₁₆H₂₁N₃O₃,xHCl
  • 1065254.
    Bendamustine Related Compound C
    Bendamustine Related Compound C
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 3 390,00 €
    3 390,00 €
    1 * 20 mg
       3 390,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1065254.
    Description
    Bendamustine Related Compound C
    Synonym(s)
    Ethyl 4-{5-[bis(2-hydroxyethyl)amino]-1-methyl-1H -benzimidazol-2-yl}butanoate
    CAS No.
    3543-74-6
    Formula
  • 1065265.
    Bendamustine Related Compound D
    Bendamustine Related Compound D
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 210,00 €
    1 210,00 €
    1 * 20 mg
       1 210,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1065265.
    Description
    Bendamustine Related Compound D
    Synonym(s)
    4-{5-[(2-Chloroethyl)amino]-1-methyl-1H -benzimidazol-2-yl}butanoic acid hydrochloride
    CAS No.
    1797881-48-1
    Formula
    C₁₄H₁₉Cl₂N₃O₂
  • 1065276.
    Bendamustine Related Compound E
    Bendamustine Related Compound E
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 280,00 €
    1 280,00 €
    1 * 20 mg
       1 280,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1065276.
    Description
    Bendamustine Related Compound E
    Synonym(s)
    4-{5-[(2-Chloroethyl)(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoic acid
    CAS No.
    109882-27-1
    Formula
    C₁₆H₂₂ClN₃O₃
  • 1065298.
    Bendamustine Related Compound G
    Bendamustine Related Compound G
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 280,00 €
    1 280,00 €
    1 * 20 mg
       1 280,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1065298.
    Description
    Bendamustine Related Compound G
    Synonym(s)
    4-[6-(2-Chloroethyl)-3,6,7,8-tetrahydro-3-methylimidazo[4,5-h][1,4]benzothiazin-2-yl]butanoic acid hydrochloride
    CAS No.
    Formula
  • 1065312.
    Bendamustine Related Compound I
    Bendamustine Related Compound I
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 110,00 €
    1 110,00 €
    1 * 25 mg
       1 110,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1065312.
    Description
    Bendamustine Related Compound I
    Synonym(s)
    Ethyl 4-{5-[bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoate
    CAS No.
    87475-54-5
    Formula
    C₁₈H₂₅Cl₂N₃O₂