Atorvastatin Methyl Ester, Pharmaceutical Analytical Impurities
Atorvastatin Methyl Ester, Pharmaceutical Analytical Impurities, Vial, 25 mg
1A00020.
: USP
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- Description:Atorvastatin Methyl Ester
- Cat. No.:1A00020.
- Description:Atorvastatin Methyl Ester
- Synonym(s):methyl (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate
- CAS No.:345891-62-5



