Order Entry
France
Orders LinkContactUsLinkComponent
Triptorelin Related Compounds, Reference Standard

Triptorelin Related Compounds, Reference Standard

Supplier:   USP
Triptorelin Related Compounds, Reference Standard
Supplier:  USP
Certificates
Triptorelin Related Compounds, Reference Standard

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1696110.
  • 1696128.
  • 1696131.
  • 1696142.
  • Description:
  • Triptorelin Related Compound A
  • Triptorelin Related Compound B
  • Triptorelin Acetate
  • Triptorelin Related Compound C
  • Synonym(s):
  • 5-Oxo-L-prolyl-L-histidyl-L-tryptophyl-L-seryl-L-tyrosyl-D-tryptophyl-L-leucyl-L-arginyl-L-prolylglycinamide acetate salt
  • Triptorelin acid; 5-Oxo-L-prolyl-L-histidyl-L-tryptophyl-L-seryl-L-tyrosyl-D-tryptophyl-L-leucyl-L-arginyl-L-prolylglycine
  • CAS No.:
  • 102992-39-2
  • 321709-35-7
  • 57773-63-4
  • 129418-54-8

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1696110.
    Triptorelin Related Compound A
    Triptorelin Related Compound A
    USP reference standard
    Vial
    1,23 mg
    Log in for availability
    Log in for availability
    1 * 1,23 mg
    1 * 1,23 mg - 1 120,00 €
    1 120,00 €
    1 * 1,23 mg
       1 120,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1696110.
    Description
    Triptorelin Related Compound A
    Synonym(s)
    CAS No.
    102992-39-2
  • 1696128.
    Triptorelin Related Compound B
    Triptorelin Related Compound B
    USP reference standard
    Vial
    1,68 mg
    Log in for availability
    Log in for availability
    1 * 1,68 mg
    1 * 1,68 mg - 1 120,00 €
    1 120,00 €
    1 * 1,68 mg
       1 120,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1696128.
    Description
    Triptorelin Related Compound B
    Synonym(s)
    CAS No.
    321709-35-7
  • 1696131.
    Triptorelin Acetate
    Triptorelin Acetate
    USP reference standard
    Vial
    1,82 mg
    Log in for availability
    Log in for availability
    1 * 1,82 mg
    1 * 1,82 mg - 455,00 €
    455,00 €
    1 * 1,82 mg
       455,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1696131.
    Description
    Triptorelin Acetate
    Synonym(s)
    5-Oxo-L-prolyl-L-histidyl-L-tryptophyl-L-seryl-L-tyrosyl-D-tryptophyl-L-leucyl-L-arginyl-L-prolylglycinamide acetate salt
    CAS No.
    57773-63-4
  • 1696142.
    Triptorelin Related Compound C
    Triptorelin Related Compound C
    USP reference standard
    Vial
    1,31 mg
    Log in for availability
    Log in for availability
    1 * 1,31 mg
    1 * 1,31 mg - 1 180,00 €
    1 180,00 €
    1 * 1,31 mg
       1 180,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1696142.
    Description
    Triptorelin Related Compound C
    Synonym(s)
    Triptorelin acid; 5-Oxo-L-prolyl-L-histidyl-L-tryptophyl-L-seryl-L-tyrosyl-D-tryptophyl-L-leucyl-L-arginyl-L-prolylglycine
    CAS No.
    129418-54-8
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.