Order Entry
France
Orders LinkContactUsLinkComponent
Trifluridine, Reference Standards

Trifluridine, Reference Standards

Supplier:   USP
Trifluridine, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1686309.
  • 1686310.
  • Description:
  • Trifluridine
  • Trifluridine Related Compound A
  • Synonym(s):
  • 5-Carboxy-2'-deoxyuridine
  • CAS No.:
  • 70-00-8
  • 14599-46-3

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1686309.
    Trifluridine
    Trifluridine
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 518,00 €
    518,00 €
    1 * 100 mg
       518,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1686309.
    Description
    Trifluridine
    Synonym(s)
    CAS No.
    70-00-8
  • 1686310.
    Trifluridine Related Compound A
    Trifluridine Related Compound A
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 300,00 €
    1 300,00 €
    1 * 10 mg
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1686310.
    Description
    Trifluridine Related Compound A
    Synonym(s)
    5-Carboxy-2'-deoxyuridine
    CAS No.
    14599-46-3