Riociguat, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Riociguat System Suitability Mixture: This is a mixture containing (approximately 0.2% of related compound B and 0.5% of related compound C) riociguat, riociguat related compound B and riociguat related compound C
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Riociguat System Suitability Mixture: This is a mixture containing (approximately 0.2% of related compound B and 0.5% of related compound C) riociguat, riociguat related compound B and riociguat related compound C
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1604494.
- 1604519.
- 1604527.
- Description:
- Riociguat System Suitability Mixture II
- Riociguat
- Riociguat System Suitability Mixture
- CAS No.:
- 625115-55-1
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1604494.Riociguat System Suitability Mixture IIRiociguat System Suitability Mixture IIUSP reference standardVial25 mgLog in for availability1 * 25 mg1 230,00 €Specifications:
More
Specifications
Cat. No.1604494.DescriptionRiociguat System Suitability Mixture IICAS No. - 1604519.RiociguatSpecifications:
More
Specifications
Cat. No.1604519.DescriptionRiociguatCAS No.625115-55-1 - 1604527.Riociguat System Suitability MixtureRiociguat System Suitability MixtureUSP reference standardVial25 mgLog in for availability1 * 25 mg1 330,00 €Specifications:
More
Specifications
Cat. No.1604527.DescriptionRiociguat System Suitability MixtureCAS No.