Order Entry
France
Orders LinkContactUsLinkComponent
Nilotinib, Reference Standards

Nilotinib, Reference Standards

Supplier:   USP
Nilotinib, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1463428.
  • 1463461.
  • Description:
  • Nilotinib
  • Nilotinib System Suitability Mixture
  • CAS No.:
  • 923288-90-8
  • UN No.:
  • 3077
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1463428.
    Nilotinib
    Nilotinib
    USP reference standard
    Vial
    350 mg
    Log in for availability
    Log in for availability
    1 * 350 mg
    1 * 350 mg - 722,00 €
    722,00 €
    1 * 350 mg
       722,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1463428.
    Description
    Nilotinib
    CAS No.
    923288-90-8
    UN No.
    3077
  • 1463461.
    Nilotinib System Suitability Mixture
    Nilotinib System Suitability Mixture
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 260,00 €
    1 260,00 €
    1 * 10 mg
       1 260,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1463461.
    Description
    Nilotinib System Suitability Mixture
    CAS No.
    UN No.
    3077