Order Entry
France
Orders LinkContactUsLinkComponent
 

Nitroglycerin, Reference Standards

 :   USP
Nitroglycerin, Reference Standards
 :  USP
 
Nitroglycerin, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

 

  • Catalog No:
  • 1213148.
  • 1213159.
  • 1445571.
  • 1445584.
  • Description:
  • 1,2-Dinitroglycerin
  • 1,3-Dinitroglycerin
  • 1-Mononitroglycerin
  • 2-Mononitroglycerin
  • CAS No.:
  • 621-65-8
  • 623-87-0
  • 624-43-1
  • 620-12-2
  • UN No.:
  • 1648
  • 1648
  • 1648
  • 1648

 

Product Family Options

 

  • Description
    Standards
    Pk Info
    Pack type
    Pk
     
     
  • 1,2-Dinitroglycerin
    USP reference standard
    2 ampules x 1 mL
    Ampule
    2 mL
       
       
    1 * 2 mL
    1 * 2 mL - 1 170,00 €
    1 170,00 €
    1 * 2 mL
       1 170,00 €

     
     
    Specifications
    Cat. No.
    1213148.
    Description
    1,2-Dinitroglycerin
    CAS No.
    621-65-8
    UN No.
    1648
  • 1,3-Dinitroglycerin
    USP reference standard
    2 ampules x 1 mL
    Ampule
    2 mL
       
       
    1 * 2 mL
    1 * 2 mL - 1 170,00 €
    1 170,00 €
    1 * 2 mL
       1 170,00 €

     
     
    Specifications
    Cat. No.
    1213159.
    Description
    1,3-Dinitroglycerin
    CAS No.
    623-87-0
    UN No.
    1648
  • 1-Mononitroglycerin
    USP reference standard
    2 ampules x 1 mL
    Ampule
    2 mL
       
       
    1 * 2 mL
    1 * 2 mL - 1 170,00 €
    1 170,00 €
    1 * 2 mL
       1 170,00 €

     
     
    Specifications
    Cat. No.
    1445571.
    Description
    1-Mononitroglycerin
    CAS No.
    624-43-1
    UN No.
    1648
  • 2-Mononitroglycerin
    USP reference standard
    2 ampules x 1 mL
    Ampule
    2 mL
       
       
    1 * 2 mL
    1 * 2 mL - 1 130,00 €
    1 130,00 €
    1 * 2 mL
       1 130,00 €

     
     
    Specifications
    Cat. No.
    1445584.
    Description
    2-Mononitroglycerin
    CAS No.
    620-12-2
    UN No.
    1648