Order Entry
France
Orders LinkContactUsLinkComponent
 

Fixed-Angle Titanium Rotor, Type 70.1 Ti

Type 70.1 Ti fixed angle titanium rotor. For use in instruments classified: HRS. Major applications: Differential centrifugation of subcellular particles, Fixed angle (ultracentrifugation)
  521-3729
Fixed-Angle Titanium Rotor, Type 70.1 Ti
  521-3729
Fixed-Angle Titanium Rotor, Type 70.1 Ti
Fixed-Angle Titanium Rotor, Type 70.1 Ti

Type 70.1 Ti fixed angle titanium rotor. For use in instruments classified: HRS. Major applications: Differential centrifugation of subcellular particles, Fixed angle (ultracentrifugation)

 
25 082,00 €
   
1 * 1 Pce
 

Type 70.1 Ti Fixed angle titanium rotor.

For use in instruments classified: HRS. Major applications: Differential centrifugation of subcellular particles.

 
  • Description:
    Type 70.1 Ti fixed angle titanium rotor. For use in instruments classified: HRS. Major applications: Differential centrifugation of subcellular particles
  • For:
    Fixed angle (ultracentrifugation)
  • Cat. No.:
    521-3729
  • Radius:
    82 mm
  • No. of tubes accepted:
    12×14 ml/16×76 mm (12×14 ml/1×3")
  • Max. RPM:
    70000