Silica gel, white, small, pored beads 2,5 - 4 mm
Catalog # 27612.292
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Bulk density:680 - 780 g/L
- Adsorption capacity (H₂O; 23 °C; 80% RH):Min. 21,5%
- Moisture (loss on drying (140 °C):Max. 2,0%
Specification Test Results
About this item
:
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa) (lit.)
Melting Pt:> 1600 °C (lit.)
Storage Temperature:Ambient
CAS Number:7631-86-9
:63231-67-4
Formula:SiO₂
MDL Number:MFCD00011232