TMEM173/MPYS overexpression lysate (adult normal)
Catalog # NOVUNBL1-17038
Supplier: Novus Biologicals
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Pk:0,1 mg
- Tested applications:WB
Specifications
About this item
TMEM173/MPYS Overexpression Lysate (Adult Normal)
Tested Applications: Western Blot.