Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Forceps
Dumont SS fine forceps
Dumont SS fine forceps
Dumont SS fine forceps
Catalog # FSCI11200-33
Supplier:
FINE SCIENCE TOOLS
Expand image
Dumont SS fine forceps
Catalog # FSCI11200-33
Supplier:
FINE SCIENCE TOOLS
Specifications
Type:
SS
Length:
135 mm
Material:
Dumoxel
Tip style:
Biology
Pk:
1
Tip size:
0,1×0,06 mm
Tip shape:
Straight
Specifications
About this item
Product Details
Product Details