VWR® Media Supplements for Dehydrated Culture Media
Supplier: VWR Chemicals
Certificates
About this item
These standard supplements can be used with several culture media to prepare the complete culture media for your specific purpose. Most of these supplements are available as freeze-dried in vials and this ensures.
- No toxic agent aerosols released upon opening
- Standard volumes make preparation easier
- User friendly screw caps
- Can be stored at room temperature
Related Information
Certificates
Search by Lot Number303570ZK_Polysorbato 80_ISO 11133_rev1 335032ZF_LISTERIA ENRICHMENT SUPLEMENT( Ottaviani & Agosti) 10x24_ISO 11133_rev1 355070ZF_ROSOLIC ACID SOLUTION 1% 100ML_ISO 11133_rev1 602730ZL_RPF SUPPLEMENT 10X10ML_ISO 11133_rev1 928010NL_AMPICILLIN SELECTIVE SUPPLEMENT_ISO 11133_rev1
More
SDS_EN_PL_VWRC335032ZF.pdf SDS_EN_UK_VWRC303570.pdf SDS_EN_UK_VWRC335032ZF.pdf SDS_EN_UK_VWRC602730ZL.pdf SDS_EN_UK_VWRC928180NL.pdf
SDS
More
spec_VWRC303570ZK spec_VWRC335032ZF spec_VWRC355070ZF spec_VWRC602730ZL spec_VWRC928010NL
Product specification
More
Frequently Bought Together