HyClone™ Donor Equine Serum, U.S. origin, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
Certificates
About this item
The animals are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration. The horses are bled in an enclosed facility that is specially equipped for aseptic collection. Coggins tests for equine infectious anemia are routinely performed on each horse.
- Tested for equine infectious anemia
- Complete traceability back to original source
- Virus panel testing according to 9 CFR 113,53
onor Equine Serum is filtered through three sequential 100 nm (0,1 μm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
Specifications
- Catalog No:
- SH30074.04IR25
- HYCLSH30074.04
- Origin:
- US
- US
- Endotoxin:
- ≤10 EU/ml
- ≤10 EU/ml
- Haemoglobin:
- ≤10 mg/dl
- ≤10 mg/dl
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
Specifications
Frequently Bought Together