Order Entry
France
ContactUsLinkComponent
Lambda gt11 vector kits
Lambda gt11 vector kits
Catalog # STRA234211
Supplier:  STRATAGENE
CAS Number:  
undefined
Lambda gt11 vector kits
Catalog # STRA234211
Supplier:  STRATAGENE
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Lambda gt11/EcoR I/CIAP Treated Vector Kit, vector arms dephosphorylated with alkaline phosphatase to prevent self-ligation. Contains: 10 μg lambda gt11/EcoR I/CIAP-treated vector, host strains: Y1088, Y1089r-, Y1090r-, test insert
  • Seq:
    01
Specifications
About this item
For use in the construction of cDNA libraries.
  • Predigested with EcoR I
  • >1×10⁷ pfu/μg lambda DNA must be obtained with test insert (background <1,5%)
These Lambda gt11 vector kits do not allow in vivo excision of cloned fragments. Libraries may be screened with both antibodies and nucleic acid probes.
Caution: For research use only. Not for use in diagnostic procedures.