Transfection reagents, GeneJammer
Supplier: AGILENT
Certificates
About this item
The GeneJammer transfection reagent is a proprietary formulation of novel polyamines ideal for transient or stable transfections in a wide variety of cell types.
- Fast and easy to use, no media changes required
- Enables high level adenovirus transduction in the absence of an adenovirus specific receptor
- Works with media containing antibiotics, growth factors and/or other supplements
This transfection reagent is effective in both serum-containing medium and serum-free medium, offering high efficiencies with minimal cytotoxicity.
1.33 mg/ml reagent in 80% ethanol.
Caution: For research use only. Not for use in diagnostic procedures.
Frequently Bought Together