Order Entry
France
ContactUsLinkComponent
 
J.T.Baker® BAKER INSTRA-ANALYZED®, Potassium Plasma Standard
  5774.0100
 :  Avantor
 :  
J.T.Baker® BAKER INSTRA-ANALYZED®, Potassium Plasma Standard
  5774.0100
 :  Avantor
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Element:
    Potassium
  • Symbol:
    K