0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Antibodies
Anti-CD20 Antibody
Anti-CD20 Antibody
APOSBITP2047-1MG
:
Apollo Scientific
Anti-CD20 Antibody
APOSBITP2047-1MG
:
Apollo Scientific
Antigen symbol:
CD20
Reactivity:
Human
Pk:
1 mg