Order Entry
France
ContactUsLinkComponent
Anti-CD4 Rat Antibody
Anti-CD4 Rat Antibody
Catalog # APOSBITP2020-1MG
Supplier:  Apollo Scientific
Anti-CD4 Rat Antibody
Catalog # APOSBITP2020-1MG
Supplier:  Apollo Scientific

Specifications

  • Antigen symbol:
    CD4
  • Host:
    Rat
  • Isotype:
  • Reactivity:
    Mouse
  • Environmentally Preferable:
  • Pk:
    1 mg

Specifications

About this item