Order Entry
France
ContactUsLinkComponent
Tagged column package, His 2-Step purification protocol
Tagged column package, His 2-Step purification protocol
Catalog # 29-4976-30
Supplier:  Cytiva
undefined
Tagged column package, His 2-Step purification protocol
Catalog # 29-4976-30
Supplier:  Cytiva

Specifications

  • Description:
    Tagged column package, His (2-step purification protocol)
  • Storage:
    4...30 °C
  • Pk:
    1 KIT
  • Sample volume:
    5 ml

Specifications

About this item

Column package includes affinity chromatography and size exclusion chromatography columns to achieve very high purity of his-tagged proteins using a 2-step purification protocol.

  • Contains a HisTrap HP column 1 ml prepacked with Ni Sepharose High Performance (HP) affinity resin for preparative purification of his-tagged proteins
  • Final purity achieved with HiPrep 16/60 Sephacryl S-200 HR size exclusion chromatography column, removes the trace of impurities or closely related substances and imidazole
  • Includes instructions with guidance and tips showing how to get started

Ni Sepharose HP affinity resin consists of highly cross-linked agarose beads to which a chelating group has been coupled. This chelating group is precharged with nickel, which selectively retains proteins with exposed histidine groups. Ni Sepharose HP has a binding capacity that exceeds 40 mg/ml, resulting in high protein recovery.

The size exclusion column is prepacked with Sephacryl S-200 HR chromatography resin, which has a broad fractionation range of Mr 5000 to ~ 250000 for globular proteins. The high sample capacity of the HiPrep column.